Learn More
Biolegend PerCP/Cyanine5.5 anti-ZAP70 Phospho (Tyr319)/Syk Phospho (Tyr352)
Supplier:  Biolegend 683709
Encompass_Preferred
PerCP/Cyanine5.5 anti-ZAP70 Phospho (Tyr319)/Syk Phospho (Tyr352) [1503310]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-7481