Learn More
Abcam PerCP Conjugation Kit - Lightning-Link, 3 x 100UG
Catalog No. 502585586
Encompass_Preferred
PerCP conjugation / labeling in < 4 hrs with 30 secs hands-on time using PerCP Conjugation Kit - Lightning-Link ab102907. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5586 Supplier Abcam Supplier No. AB1029073X100UG
PerCP conjugation / labeling in < 4 hrs with 30 secs hands-on time using PerCP Conjugation Kit - Lightning-Link ab102907. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement