Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD11c
Supplier:  Biolegend 301623
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-165-729