Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 352309
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD138 (Syndecan-1) [DL-101]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-168-712