Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD14
Supplier:  Biolegend 301824
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD14 [M5E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Capuchin Monkey, Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Dog (Canine), Rhesus, Pigtailed Macaque, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-165-766