Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD16
Supplier:  Biolegend 302027
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-165-800