Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD1c
Supplier:  Biolegend 331513
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-661