Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD206 (MMR)
Supplier:  Biolegend 321121
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD206 (MMR) [15-2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-072