Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD21
Supplier:  Biolegend 354908
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD21 [Bu32]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-403-520