Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD3
Supplier:  Biolegend 300328
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD3 [HIT3a]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-165-549