Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD38
Supplier:  Biolegend 303521
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC; Size: 25 tests
Catalog No. 50-166-009