Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD45RA
Supplier:  Biolegend 304121
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-166-084