Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD68
Supplier:  Biolegend 333813
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-167-749