Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD69
Supplier:  Biolegend 310925
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD69 [FN50]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-597