Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD71
Supplier:  Biolegend 334114
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-071