Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD8a
Supplier:  Biolegend 301032
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. 50-165-668