Learn More
Biolegend PerCP/Cyanine5.5 anti-human IL-17A
Supplier:  Biolegend 512314
Encompass_Preferred
PerCP/Cyanine5.5 anti-human IL-17A [BL168]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-170-129