Learn More
Biolegend PerCP/Cyanine5.5 anti-human TIGIT (VSTM3)
Supplier:  Biolegend 372718
Encompass_Preferred
PerCP/Cyanine5.5 anti-human TIGIT (VSTM3) [A15153G]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1411423