Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD357 (GITR)
Supplier:  Biolegend 371218
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD357 (GITR) [108-17]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1438396