Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD45
Supplier:  Biolegend 368504
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1255817