Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD80
Supplier:  Biolegend 305232
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD80 [2D10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1475410