Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD154
Supplier:  Biolegend 106514
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD154 [MR1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-711-840