Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD103
Supplier:  Biolegend 121416
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9914980