Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD134 (OX-40)
Supplier:  Biolegend 119425
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD134 (OX-40) [OX-86]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1394430