Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD223 (LAG-3)
Supplier:  Biolegend 125211
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1022789