Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD226 (DNAM-1)
Supplier:  Biolegend 128814
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD226 (DNAM-1) [10E5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1181371