Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD279 (PD-1)
Supplier:  Biolegend 109120
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD279 (PD-1) [RMP1-30]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1057157