Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD106
Supplier:  Biolegend 105716
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD106 [429 (MVCAM.A)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-078