Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD11a/CD18 (LFA-1)
Supplier:  Biolegend 141007
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD11a/CD18 (LFA-1) [H155-78]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-175