Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD14
Supplier:  Biolegend 123313
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD14 [Sa14-2]; Isotype: Rat IgG2a, κ; Reactivity: mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-090