Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD150 (SLAM)
Supplier:  Biolegend 115922
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD150 (SLAM) [TC15-12F12.2]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-592