Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD16/32
Supplier:  Biolegend 101324
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD16/32 [93]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-552