Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD19
Supplier:  Biolegend 152406
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD19 [1D3/CD19]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-113-0314