Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD25
Supplier:  Biolegend 101912
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD25 [3C7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-613