Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD28
Supplier:  Biolegend 102114
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-648