Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD45.1
Supplier:  Biolegend 110728
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-432