Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD45
Supplier:  Biolegend 103131
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-793