Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD69
Supplier:  Biolegend 104522
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-951