Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CX3CR1
Supplier:  Biolegend 149010
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CX3CR1 [SA011F11]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-850-441