Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse H-2Kb
Supplier:  Biolegend 116515
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype; Apps: FC; Size: 25 ug
Catalog No. 50-163-674