Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse TNF-α
Supplier:  Biolegend 506321
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-169-966