Learn More
Biolegend PerCP/Cyanine5.5 anti-rat CD45RA
Catalog No. NC1285581
Encompass_Preferred
PerCP/Cyanine5.5 anti-rat CD45RA [OX-33]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 100 ug
This item is not returnable. View return policy
Catalog No. NC1285581 Supplier Biolegend Supplier No. 202318
Add to Cart
Add to Cart
PerCP/Cyanine5.5 anti-rat CD45RA [OX-33]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 100 ug