Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD160
Supplier:  Biolegend 341210
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD160 [BY55]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1546278