Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD73
Supplier:  Biolegend 127214
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD73 [TY/11.8]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1083759