Learn More
Cytek Biosciences Inc PerCP-Cy5.5 aHu CD14 Anti
Supplier: Cytek Biosciences Inc 650149T100
Product Name: PerCP-Cyanine5.5 Anti-Human CD14 (61D3)
Product Class: Antibody
Type: Primary
Product Name: PerCP-Cyanine5.5 Anti-Human CD14 (61D3)
Cytek Catalog Number: 65-0149-T100
Marker: CD14
Target Species: Human
Clonality: Monoclonal
Clone: 61D3
Host Species: Mouse
Isotype: IgG1, k
Conjugation: PerCP-Cyanine5.5
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.