Learn More
Cytek Biosciences Inc PERCP-CY5.5 HU CD3 (UCHT1)100T
Supplier:  Cytek Biosciences Inc 650038T100
Encompass_Preferred
PerCP-Cyanine5.5 Anti-Human CD3 (UCHT1) Isotype: Mouse IgG1, kappa Apps: FC Size: 100 tests
Catalog No. 50-105-4915