Learn More
Cytek Biosciences Inc PERCP-CY5.5 MS CD4(RM4-5)100UG
Supplier:  Cytek Biosciences Inc 650042U100
Encompass_Preferred
PerCP-Cyanine5.5 Anti-Mouse CD4 (RM4-5) Isotype: Rat IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-4919