Learn More
Biolegend PerCP/Cyanine5.5 Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400338
Encompass_Preferred
PerCP/Cyanine5.5 Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-168-915