Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD195 (CCR5)
Supplier:  Biolegend 359112
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-7803