Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD243 (MDR-1)
Supplier:  Biolegend 348611
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD243 (MDR-1) [UIC2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests
Catalog No. 50-207-6892